Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200
Thermo Scientific™
Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호색상멸균형식제품 사이즈
T070RLNS-QNaturalSterileRefillEach
T070-QNaturalNon-sterileBaggedCase of 10000
T070RLN-QNaturalNon-sterileRefillCase of 4800
T070RNS-QClearSterileHinged RackCase of 4800
T070RNS-QPK
T070RNS-Q으로도 사용됨
ClearSterileHinged RackPack of 960
T090-QYellowNon-sterileBaggedCase of 10000
T090R-QYellowNon-sterileHinged RackCase of 4800
T090R-QPK
T090R-Q으로도 사용됨
YellowNon-sterileHinged RackPack of 960
T090RL-QYellowNon-sterileRefillCase of 4800
T090RL-QPK
T090RL-Q으로도 사용됨
YellowNon-sterileRefillPack of 960
T090RLS-QYellowSterileRefillCase of 4800
T090RLS-QPK
T090RLS-Q으로도 사용됨
YellowSterileRefillPack of 960
T090RS-QYellowSterileHinged RackCase of 4800
T090RS-QPK
T090RS-Q으로도 사용됨
YellowSterileHinged RackPack of 960
카탈로그 번호 T070RLNS-Q
제품 가격(KRW)
250,000
온라인 행사
Ends: 30-Sep-2026
294,000
할인액 44,000 (15%)
Each
색상:
Natural
멸균:
Sterile
형식:
Refill
Unit Size:
Each
제품 가격(KRW)
250,000
온라인 행사
Ends: 30-Sep-2026
294,000
할인액 44,000 (15%)
Each
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
Featuring:
  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
사양
클린 클레임DNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
설명QSP 200, Non-Filtered
눈금이 있음Yes
길이(미터법)5.1 cm
색상Natural
필터링됨Non-filtered
용도 (장비)Universal Fit
형식Refill
물질Virgin Polypropylene
제품라인QSP
멸균Sterile
팁 스타일Beveled
부피(미터법)1 to 200 μL
Unit SizeEach