Thermo Scientific™ QSP™ Soft Collar Pipette Tip
Thermo Scientific™
Thermo Scientific™ QSP™ Soft Collar Pipette Tip
Thermo Scientific™ QSP™ Soft Collar Pipette Tips designed for multi-channel pipetting delivering even aspirations and dispenses.
Have Questions?
Catalog NumberPackaging
T070RN-Q4800/Cs.
Catalog number T070RN-Q
Price (MXN)
-
Packaging:
4800/Cs.
Thermo Scientific™ QSP™ Soft Collar natural color Pipette Tip 200μl, packaged in rack non-sterile with clean claims; Rnase/Dnase, DNA, pyrogen, and endotoxin free. Universal fit on most pipettes.
  • Soft Collar designed to work with multi-channel pipettes
  • Soft collar helps to seat pipette tips securely and evenly, offering uniform liquid aspiration and dispensing across all pipette channels
  • Universal fitting tips fit on a broad range of pipettes
Specifications
Clean ClaimsNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
Packaging4800/Cs.
AutoclavableAutoclavable
ColorNatural
For Use With (Equipment)Universal Fit
Material100% Virgin Polypropylene
Product LineQSP
SterilityNon-sterile
Tip StyleUniversal Fit
Volume (Metric)200 μL
Unit SizeCase of 4800