Micro Length Non-Filtered Pipette Tips, 0.1-10 μL
Thermo Scientific™
Micro Length Non-Filtered Pipette Tips, 0.1-10 μL
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoFormatoEsterilidad
T104RL-QRecargaNo estéril
104-QBaggedNo estéril
T104R-QHinged RackNo estéril
T104RLS-QRecargaEstéril
T104RS-QHinged RackEstéril
Número de catálogo T104RL-Q
Precio (MXN)
-
Formato:
Recarga
Esterilidad:
No estéril
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
Featuring:
  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Especificaciones
Declaraciones de limpiezaLibre de ADNasa, ARNasa, DNA y pirógenos
DescripciónQSP 10, sin filtrado
GraduadoSí
Características ecológicasACT Ecolabel (Silver)
Longitud (métrico)3.1 cm
ColorNatural
filtradas porSin filtro
Para utilizar con (equipo)Universal Fit
FormatoRecarga
MaterialPolipropileno virgen
Línea de productosQSP
EsterilidadNo estéril
Estilo de puntaMicroPoint
Volumen (métrico)10 μL
Unit SizeCase of 4800