Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200
Thermo Scientific™

Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColorEsterilidadFormato
T110BRLS-QYellowEstérilRecarga
T070-QNaturalNo estérilBagged
T070RLN-QNaturalNo estérilRecarga
T070RLNS-QNaturalEstérilRecarga
T070RNS-QClearEstérilHinged Rack
T090-QYellowNo estérilBagged
T090R-QYellowNo estérilHinged Rack
T090RL-QYellowNo estérilRecarga
T090RLS-QYellowEstérilRecarga
T090RS-QYellowEstérilHinged Rack
T110BRS-QYellowEstérilHinged Rack
Número de catálogo T110BRLS-Q
Precio (MXN)
-
Color:
Yellow
Esterilidad:
Estéril
Formato:
Recarga
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Especificaciones
Declaraciones de limpiezaADNasa, ARNasa, ADN, ATP, inhibidor de PCR y sin pirógenos
DescripciónQSP 200, sin filtrado
GraduadoSí
Longitud (métrico)5 cm
ColorYellow
filtradas porSin filtro
Para utilizar con (equipo)Universal Fit
FormatoRecarga
MaterialPolipropileno virgen
Línea de productosQSP
EsterilidadEstéril
Estilo de puntaBiselado
Volumen (métrico)De 1 a 200 μL
Unit SizeCase of 4800