QSP Specialty Pipette Tips
Thermo Scientific™

QSP Specialty Pipette Tips

Specialty pipette tips designed for specific applications making it easier to concentrate on what is important—your research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleSterilityUnit Size
T112NXLGRS-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
103-QQSP 10 Gel, Non-Filtered10 μLGel LoadingNon-sterileCase of 10000
118-N-QQSP 200 Genomic/Wide Orifice, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 10000
T118R-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 4800
T118RS-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
TF118-200-QQSP 200 Genomic, Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
112NXLG-QQSP 1000 Genomic Wide Orifice, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 10000
T112NXLGR-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 4800
TF112G-1000
also known as TF112G-1000-Q
QSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterileCase of 5
TF112G-1000-QQSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
Catalog number T112NXLGRS-Q
Price (CLP)
-
Description:
QSP 1000 Genomic, Non-Filtered
Volume (Metric):
1000 μL
Tip Style:
Genomic/Wide Orifice
Sterility:
Sterile
Unit Size:
Case of 4800
Thermo Scientific™ QSP specialty pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Specialty tips are crafted for specific applications and are designed to make your work more efficient and reproducible.

Gel Loading Specialty Tips
Loading acrylamide and agarose gels with standard pipette tips is a lengthy and tedious process. Avoid damaging your gel by choosing a specialty tip that offers a longer taper with a thinner distal end that is designed for these applications.

Genomic Wide Orifice Specialty Tips:
A problem posed when working with viscous liquids like tween or thick cell lysates is pipetting reliability due to the inability to draw and dispense fluid in a uniform manner. Our large, wide orifice tips are much wider than standard tips improving capability to draw and dispense in a consistent and uniform manner.

Extended Length Specialty Tips:
Extended length tips help prevent contamination by keeping the pipette shaft from entering the sample vessel. Extended length tips can provide laboratory solutions for dispensing samples into hard to reach vessels were a standard tip just cant reach.

Clean Claims:
QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
DescriptionQSP 1000 Genomic, Non-Filtered
Length (Metric)8.9cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialVirgin Polypropylene
Product LineQSP
SterilitySterile
Tip StyleGenomic/Wide Orifice
Volume (Metric)1000 μL
Unit SizeCase of 4800