XL Graduated MicroPoint Pipette Tips, 100-1250 μL
Thermo Scientific™

XL Graduated MicroPoint Pipette Tips, 100-1250 μL

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFormatSterility
T112NXLR-QNaturalHinged RackNon-sterile
112NXL-QNaturalBulkNon-sterile
112XL-QBlueBulkNon-sterile
T112NXLRLS-QNaturalRefillSterile
T112NXLRS-QNaturalHinged RackSterile
T112XLR-QBlueHinged RackNon-sterile
T112XLRL-QBlueRefillNon-sterile
T112XLRLS-QBlueRefillSterile
T112XLRS-QBlueHinged RackSterile
Catalog number T112NXLR-Q
Price (MXN)
-
Color:
Natural
Format:
Hinged Rack
Sterility:
Non-sterile
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 1000, Non-Filtered
GraduatedYes
Length (Metric)8.9 cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialVirgin Polypropylene
Product LineQSP
SterilityNon-sterile
Tip StyleExtended Length
Volume (Metric)1250 μL
Unit SizeCase of 4800