XL Graduated MicroPoint Pipette Tips, 100-1250 μL
Thermo Scientific™

XL Graduated MicroPoint Pipette Tips, 100-1250 μL

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColorFormatoEsterilidadFormato
T112NXLRS-QNaturalHinged RackEstérilCase of 4800
112NXL-QNaturalBulkNo estérilCase of 10000
112XL-QAzulBulkNo estérilCase of 10000
112XL-QPK
también denominado 112XL-Q
AzulBulkNo estérilPack of 1000
T112NXLR-QNaturalHinged RackNo estérilCase of 4800
T112NXLRLS-QNaturalRecargaEstérilCase of 4800
T112XLR-QAzulHinged RackNo estérilCase of 4800
T112XLRL-QAzulRecargaNo estérilCase of 4800
T112XLRLS-QAzulRecargaEstérilCase of 4800
T112XLRS-QAzulHinged RackEstérilCase of 4800
Número de catálogo T112NXLRS-Q
Precio (USD)
-
Color:
Natural
Formato:
Hinged Rack
Esterilidad:
Estéril
Unit Size:
Case of 4800
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Especificaciones
Declaraciones de limpiezaADNasa, ARNasa, ADN, ATP, inhibidor de PCR y sin pirógenos
DescripciónQSP 1000, sin filtrado
GraduadoSí
Longitud (métrico)8.9 cm
ColorNatural
filtradas porSin filtro
Para utilizar con (equipo)Universal Fit
FormatoHinged Rack
MaterialPolipropileno virgen
Línea de productosQSP
EsterilidadEstéril
Estilo de puntaLongitud extendida
Volumen (métrico)1250 μL
Unit SizeCase of 4800