QSP Filter and Non-Filtered Pipette Tips
Thermo Scientific™

QSP Filter and Non-Filtered Pipette Tips

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명부피(미터법)팁 스타일멸균제품 사이즈
T112XLRL-QPK
T112XLRL-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterilePack of 960
095-QQSP 5000, Non-Filtered5000 μLMacroNon-sterileCase of 2500
095-QPK
095-Q으로도 사용됨
QSP 5000, Non-Filtered5000 μLMacroNon-sterilePack of 250
096-QQSP 5000, Non-Filtered5000 μLMacroNon-sterileCase of 2500
096-QPK
096-Q으로도 사용됨
QSP 5000, Non-Filtered5000 μLMacroNon-sterilePack of 250
097-QQSP 10ML, Non-Filtered10 mLMacroNon-sterileCase of 2000
097-QPK
097-Q으로도 사용됨
QSP 10ML, Non-Filtered10 mLMacroNon-sterilePack of 200
090-QQSP 5000, Non-Filtered5000 μLMacroNon-sterileCase of 2500
090-QPK
090-Q으로도 사용됨
QSP 5000, Non-Filtered5000 μLMacroNon-sterilePack of 250
104-QQSP 10, Non-Filtered10 μLMicroPointNon-sterileCase of 10000
104-QPK
104-Q으로도 사용됨
QSP 10, Non-Filtered10 μLMicroPointNon-sterilePack of 1000
106-QQSP 300, Non-Filtered300 μLMicroPointNon-sterileCase of 10000
106-QPK
106-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-sterilePack of 1000
102-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-sterileCase of 10000
102-QPK
102-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthNon-sterilePack of 1000
110-NXL-QQSP 200XL, Non-Filtered200 μLExtended LengthNon-sterileCase of 5000
110-NXL-QPK
110-NXL-Q으로도 사용됨
QSP 200XL, Non-Filtered200 μLExtended LengthNon-sterilePack of 1000
111-QQSP 1000, Non-Filtered1000 μLMicroPointNon-sterileCase of 10000
111-QPK
111-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLMicroPointNon-sterilePack of 1000
112NXL-QQSP 1000, Non-Filtered1250 μLExtended LengthNon-sterileCase of 10000
112NXL-QPK
112NXL-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterilePack of 1000
111-N-QQSP 1000, Non-Filtered1000 μLMicroPointNon-sterileCase of 10000
111-N-QPK
111-N-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLMicroPointNon-sterilePack of 1000
114-QQSP 20, Non-Filtered20 μLMicroPointNon-sterileCase of 10000
114-QPK
114-Q으로도 사용됨
QSP 20, Non-Filtered20 μLMicroPointNon-sterilePack of 1000
112XL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-sterileCase of 10000
112XL-QPK
112XL-Q으로도 사용됨
QSP 1000, Non-Filtered1000 μLExtended LengthNon-sterilePack of 1000
T070-QQSP 200, Non-Filtered200 μLBeveledNon-sterileCase of 10000
T070RLNS-QQSP 200, Non-Filtered1 to 200 μLBeveledSterileEach
T070RNS-QQSP 200, Non-Filtered200 μLBeveledSterileCase of 4800
T070RNS-QPK
T070RNS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledSterilePack of 960
T070RLN-QQSP 200, Non-Filtered1 to 200 μLBeveledNon-sterileCase of 4800
T090-QQSP 200, Non-Filtered200 μLBeveledNon-sterileCase of 10000
T090RL-QQSP 200, Non-Filtered200 μLBeveledNon-sterileCase of 4800
T090RL-QPK
T090RL-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-sterilePack of 960
T090RLS-QQSP 200, Non-Filtered200 μLBeveledSterileCase of 4800
T090RLS-QPK
T090RLS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledSterilePack of 960
T102R-QQSP 10XL, Non-Filtered0.1 to 10 μLExtended LengthNon-sterileCase of 4800
T102R-QPK
T102R-Q으로도 사용됨
QSP 10XL, Non-Filtered0.1 to 10 μLExtended LengthNon-sterilePack of 960
T102RS-QQSP 10XL, Non-Filtered10 μLExtended LengthSterileCase of 4800
T102RS-QPK
T102RS-Q으로도 사용됨
QSP 10XL, Non-Filtered10 μLExtended LengthSterilePack of 960
T090RS-QQSP 200, Non-Filtered200 μLBeveledSterileCase of 4800
T090RS-QPK
T090RS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledSterilePack of 960
T090R-QQSP 200, Non-Filtered200 μLBeveledNon-sterileCase of 4800
T090R-QPK
T090R-Q으로도 사용됨
QSP 200, Non-Filtered200 μLBeveledNon-sterilePack of 960
T104RL-QQSP 10, Non-Filtered10 μLMicroPointNon-sterileCase of 4800
T104RL-QPK
T104RL-Q으로도 사용됨
QSP 10, Non-Filtered10 μLMicroPointNon-sterilePack of 960
T104RLS-QQSP 10, Non-Filtered0.1 to 10 μLMicroPointSterileCase of 4800
T104RLS-QPK
T104RLS-Q으로도 사용됨
QSP 10, Non-Filtered0.1 to 10 μLMicroPointSterilePack of 960
T104R-QQSP 10, Non-Filtered10 μLMicroPointNon-sterileCase of 4800
T104R-QPK
T104R-Q으로도 사용됨
QSP 10, Non-Filtered10 μLMicroPointNon-sterilePack of 960
T104RS-QQSP 10, Non-Filtered10 μLMicroPointSterileCase of 4800
T104RS-QPK
T104RS-Q으로도 사용됨
QSP 10, Non-Filtered10 μLMicroPointSterilePack of 960
T106RL-QQSP 300, Non-Filtered300 μLMicroPointNon-sterileCase of 4800
T106RL-QPK
T106RL-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointNon-sterilePack of 960
T106RS-QQSP 300, Non-Filtered300 μLMicroPointSterileCase of 4800
T106RS-QPK
T106RS-Q으로도 사용됨
QSP 300, Non-Filtered300 μLMicroPointSterilePack of 960
T112NXLRLS-QQSP 1000, Non-Filtered1250 μLExtended LengthSterileCase of 4800
T112NXLRLS-QPK
T112NXLRLS-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthSterilePack of 960
T112NXLR-QQSP 1000, Non-Filtered1250 μLExtended LengthNon-sterileCase of 4800
T112NXLR-QPK
T112NXLR-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterilePack of 960
T112NXLRS-QQSP 1000, Non-Filtered1250 μLExtended LengthSterileCase of 4800
T112NXLRS-QPK
T112NXLRS-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthSterilePack of 960
T112XLRL-QQSP 1000, Non-Filtered1250 μLExtended LengthNon-sterileCase of 4800
T112XLRLS-QQSP 1000, Non-Filtered1250 μLExtended LengthSterileCase of 4800
T112XLRLS-QPK
T112XLRLS-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthSterilePack of 960
T112XLRS-QQSP 1000, Non-Filtered1250 μLExtended LengthSterileCase of 4800
T112XLRS-QPK
T112XLRS-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthSterilePack of 960
T112XLR-QQSP 1000, Non-Filtered1250 μLExtended LengthNon-sterileCase of 4800
T112XLR-QPK
T112XLR-Q으로도 사용됨
QSP 1000, Non-Filtered1250 μLExtended LengthNon-sterilePack of 960
TF102-10-QQSP 10XL, Filtered10 μLExtended LengthSterileCase of 4800
TF102-10-QPK
TF102-10-Q으로도 사용됨
QSP 10XL, Filtered10 μLExtended LengthSterilePack of 960
TF108-200-QQSP 200, Filtered200 μLMicroPointSterileCase of 4800
TF108-200-QPK
TF108-200-Q으로도 사용됨
QSP 200, Filtered200 μLMicroPointSterilePack of 960
TF140-200-QQSP 200, Filtered200 μLMicroPointSterileCase of 4800
TF140-200-QPK
TF140-200-Q으로도 사용됨
QSP 200, Filtered200 μLMicroPointSterilePack of 960
TF112-1000-QQSP 1000, Filtered100 to 1000 μLExtended LengthSterileCase of 4800
TF112-1000-QPK
TF112-1000-Q으로도 사용됨
QSP 1000, Filtered100 to 1000 μLExtended LengthSterilePack of 960
TF113-20-QQSP 20, Filtered20 μLBeveledSterileCase of 4800
TF113-20-QPK
TF113-20-Q으로도 사용됨
QSP 20, Filtered20 μLBeveledSterilePack of 960
TF104-10-QQSP 10, Filtered10 μLMicroPointSterileCase of 4800
TF104-10-QPK
TF104-10-Q으로도 사용됨
QSP 10, Filtered10 μLMicroPointSterilePack of 960
TTW110RNS-QQSP 200, Non-Filtered200 μLMicroPointSterileCase of 4800
TTW110RNS-QPK
TTW110RNS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLMicroPointSterilePack of 960
TF113-100-QQSP 100, Filtered100 μLBeveledSterileCase of 4800
TF113-100-QPK
TF113-100-Q으로도 사용됨
QSP 100, Filtered100 μLBeveledSterilePack of 960
TTW110RS-QQSP 200, Non-Filtered200 μLMicroPointSterileCase of 4800
TTW110RS-QPK
TTW110RS-Q으로도 사용됨
QSP 200, Non-Filtered200 μLMicroPointSterilePack of 960
TW110-N-QQSP 200, Non-Filtered200 μLMicroPointNon-sterileCase of 10000
TW110-N-QPK
TW110-N-Q으로도 사용됨
QSP 200, Non-Filtered200 μLMicroPointNon-sterilePack of 1000
카탈로그 번호 T112XLRL-QPK
T112XLRL-Q으로도 사용됨
제품 가격(KRW)
75,000
Pack of 960
설명:
QSP 1000, Non-Filtered
부피(미터법):
1250 μL
팁 스타일:
Extended Length
멸균:
Non-sterile
Unit Size:
Pack of 960
제품 가격(KRW)
75,000
Pack of 960
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:
QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

사양
클린 클레임DNase, RNase, DNA, and Pyrogen Free
설명QSP 1000, Non-Filtered
눈금이 있음Yes
길이(미터법)8.9cm
색상Blue
필터링됨Non-filtered
용도 (장비)Universal Fit
형식Refill
물질Virgin Polypropylene
제품라인QSP
멸균Non-sterile
팁 스타일Extended Length
부피(미터법)1250 μL
Unit SizePack of 960