Thermo Scientific™ QSP™ Graduation Mark Pipette Tips
El producto real puede variar
Thermo Scientific™ QSP™ Graduation Mark Pipette Tips
Thermo Scientific™ QSP™ Graduation Mark Pipette Tips
Thermo Scientific™
Thermo Scientific™ QSP™ Graduation Mark Pipette Tips
Thermo Scientific™ QSP™ Graduated Mark Pipette Tips are designed to supply a quick liquid level check, making sure you are aspirating the correct volume.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoEsterilidadColorFormatoEsterilizables en autoclave
T113R-QNo estérilAmarilloGradillaAutoclavable
T108RS-QNo estérilNaturalEn bolsaNo Autoclavable
T113RLS-QEstérilAmarilloSistema de recargaAutoclavable
T113RLN-QNo estérilNaturalSistema de recargaAutoclavable
113-G-QNo estérilAmarilloEn bolsaAutoclavable
108-G-QNo estérilNaturalEn bolsaAutoclavable
113-GN-QNo estérilNaturalEn bolsaAutoclavable
T108R-QNo estérilNaturalEn bolsaAutoclavable
T113RN-QNo estérilNaturalGradillaAutoclavable
T113RL-QNo estérilAmarilloSistema de recargaAutoclavable
T113RLNS-QEstérilNaturalSistema de recargaAutoclavable
T113RNS-QEstérilNaturalGradillaAutoclavable
T113RS-QEstérilAmarilloGradillaAutoclavable
Número de catálogo T113R-Q
Precio (MXN)
-
Esterilidad:
No estéril
Color:
Amarillo
Formato:
Gradilla
Esterilizables en autoclave:
Autoclavable
Thermo Scientific™ QSP™ Graduated Mark 200μl Natural Pipette Tips are beveled and have graduation marks at 10, 50, 100μl intervals. Packaged in bulk bags and non-sterile clean claims are; Rnase/Dnase, DNA, pyrogen and endotoxin free. Universal fit on most pipettes.
  • 200μl tip beveled and graduated at 10, 50, 100μl and available in sterile and non sterile options
  • Universal fit on broad pipette range
  • Multiple packaging options from bulk to racked and reload system.
Especificaciones
Declaraciones de limpiezaNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
GraduadoSí
Graduaciones10, 50, 100 μL
Esterilizables en autoclaveAutoclavable
ColorAmarillo
FormatoGradilla
Material100% Virgin Polypropylene
Línea de productosQSP
EsterilidadNo estéril
Estilo de puntaBeveled
Volumen (métrico)200 μL
Unit SizeCase of 4800