Luminescence Enhancer For Solution Assays, Sapphire™ Enhancer

Catalog number: T2011

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Saphire™ chemiluminescence enhancer is a concentrate reagent suitable for use with chemiluminescent substrates (AMPPD®, CSPD®, Galacton®, Glucon™, or Glucuron®).

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Format: Bottle(s)
Product Line: NovaBright™, Sapphire™
Product Size: 2500 assays
Substrate: Alkaline Phosphatase
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Room Temperature

Contents & storage

Store the product at 4°C.

Documents

Manuals & protocols