Invitrogen™

Diethanolamine (DEA)

Catalog number:  T2027

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Diethanolamine (DEA) is the buffer of choice in applications employing concentrated Tropix substrates for the detection of alkaline phosphatase. DEA buffers provide favorable reaction kinetics and greater sensitivity compared to most buffer systems. DEA is supplied 99% pure (MW = 105.1 g/mol; density = 1.097g/mL). The suggested working concentration is 0.1 M DEA (pH 10) with 1 mM MgCl2.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Products: AP Substrates
Product Line: NovaBright™
Product Size: 120 mL
Shipping Condition: Room Temperature

Contents & storage

Store the product at room temperature.

Documents

Manuals & protocols