Luminescence Enhancer For Solution Assays, Sapphire-II™ Enhancer
Invitrogen™
Luminescence Enhancer For Solution Assays, Sapphire-II™ Enhancer
Chemiluminescence enhancer that significantly reduces aqueous quenching and tune the wavelength of light emission.
Have Questions?
Catalog NumberQuantity
T21132500 assays
Catalog number T2113
Price (USD)
316.00
Each
Quantity:
2500 assays
Request bulk or custom format
Price (USD)
316.00
Each
The Sapphire-II chemiluminescence enhancer significantly reduces aqueous quenching and tune the wavelength of light emission.
  • Aqueous environments reduce chemiluminescent signal intensity of 1,2-dioxetane substrates by water-induced quenching; chemiluminescence enhancers increase the emission efficiency of light production by partitioning the water from the site of chemiluminescent signal production.
  • Chemically optimized formulations enhance signal intensity in solution-based applications with dioxetane-based substrate solutions (CSPD, CDP-Star, Galacton, Galacton-Plus, Galacton-Star, or NA-Star substrates).
  • Choice of Sapphire-II enhancer facilitates assay development by allowing users to optimize assay signal intensity and wavelength of light emission.
Optimize Chemiluminescence Signals
Sapphire-II chemiluminescence enhancer, are essential components of tube or microplate-based assays, significantly reducing the effect of aqueous quenching. Enhancers also tune the wavelength of light emission. Sapphire-II enhancer shifts the light emission maximum to 461 nm. Sapphire-II enhancer is the optimum choice for applications requiring maximum signal intensity or blue-shifted light emission.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
Product LineNovaBright, Sapphire-II
Shipping ConditionRoom Temperature
SubstrateAlkaline Phosphatase
Substrate TypeAP (Alkaline Phosphatase) Substrate
Quantity2500 assays
Unit SizeEach
Contents & Storage
Store the product at 4°C.