Invitrogen™

CSPD™ Substrate (25 mM Concentrate)

Catalog number: T2138
Invitrogen™

CSPD™ Substrate (25 mM Concentrate)

Catalog number: T2138
Catalog Number
T2138
Unit Size
100 mL
Price (USD)
Price: 13,280.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
0.5 mL
Price (USD)
Price: 189.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Price: 288.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Price: 1,038.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Price: 4,420.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Price: 2,030.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T2138100 mL
Price: 13,280.00
Your Price:
-
T20400.5 mL
Price: 189.00
Your Price:
-
T20981 mL
Price: 288.00
Your Price:
-
T20425 mL
Price: 1,038.00
Your Price:
-
T204425 mL
Price: 4,420.00
Your Price:
-
T204310 mL
Price: 2,030.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This concentrated CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
For Use With (Application)
Southern, Northern, and Western Blotting
Product Line
CSPD™, NovaBright™
Quantity
100 mL
Substrate
Alkaline Phosphatase
Substrate Type
AP (Alkaline Phosphatase) Substrate
Format
Bottle(s)
Shipping Condition
Room Temperature

Contents & Storage

Store the product at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text