Sustrato CDP-<i>Star</i>&trade; (0,25 mM listo para su uso)
Invitrogen™
Sustrato CDP-Star™ (0,25 mM listo para su uso)
Sustrato CDP-Star™ (0,25 mM listo para su uso)
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
T2146100 ml
T2147250 ml
Número de catálogo T2146
Precio (MXN)
-
Cantidad:
100 ml
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
  • Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
  • Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Método de detecciónQuimioluminiscente
FormularioLíquido
FormatoFrasco
Línea de productosCDP-Star, NovaBright
Condiciones de envíoTemperatura ambiente
Tipo de sustratoFosfatasa alcalina
Tipo de productoSustrato ELISA
Cantidad100 ml
Unit SizeEach
Contenido y almacenamiento
Almacene el producto a 4 °C.