CDP-<i>Star</i>&trade; Substrate (0.4 mM Ready-to-Use) with Enhancer
Invitrogen™
CDP-Star™ Substrate (0.4 mM Ready-to-Use) with Enhancer
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
T2214Sapphire-II™ Enhancer
T2216Emerald-II™ Enhancer
Catalog number T2214
Price (USD)
705.60
Special offer
Online exclusive
Ends: 27-Jun-2026
882.00
Save 176.40 (20%)
Each
Description:
Sapphire-II™ Enhancer
Request bulk or custom format
Price (USD)
705.60
Special offer
Online exclusive
Ends: 27-Jun-2026
882.00
Save 176.40 (20%)
Each
Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
The CDP-Star™ Substrate with Sapphire-II™ or Emerald-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancer. Membrane-based assays may also require it for increased light output and sensitivity.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionSapphire-II™ Enhancer
Detection MethodChemiluminescence
FormLiquid
FormatBottle
Product LineCDP-Star, NovaBright
Shipping ConditionRoom Temperature
Substrate TypeAlkaline Phosphatase
Product TypeELISA Substrate with Enhancer
Quantity100 mL
Unit SizeEach
Contents & Storage
Store the product at 4°C.