CDP-<i>Star</i>&trade; Substrate (0.4 mM Ready-to-Use) with Enhancer
Invitrogen™

CDP-Star™ Substrate (0.4 mM Ready-to-Use) with Enhancer

Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명
T2216Emerald-II™ Enhancer
T2214Sapphire-II™ Enhancer
카탈로그 번호 T2216
제품 가격(KRW)
1,099,000
온라인 행사
Ends: 30-Sep-2026
1,255,000
할인액 156,000 (12%)
Each
설명:
Emerald-II™ Enhancer
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
1,099,000
온라인 행사
Ends: 30-Sep-2026
1,255,000
할인액 156,000 (12%)
Each
Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

The CDP-Star™ Substrate with Sapphire-II™ or Emerald-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancer. Membrane-based assays may also require it for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.

For Research Use Only. Not for use in diagnostic procedures.
사양
설명Emerald-II™ Enhancer
검출 방법Chemiluminescence
형태Liquid
형식Bottle
제품라인CDP-Star, NovaBright
배송 조건Room Temperature
기질 유형Alkaline Phosphatase
제품 유형ELISA Substrate with Enhancer
수량100 mL
Unit SizeEach
구성 및 보관
Store the product at 4°C.