Light Emission Accelerator-II
Invitrogen™

Light Emission Accelerator-II

Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.

Have Questions?
Catalog NumberQuantity
T2222210 mL
Catalog number T2222
Price (USD)
328.00
Each
Quantity:
210 mL
Request bulk or custom format
Price (USD)
328.00
Each

Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.

For Research Use Only. Not for use in diagnostics procedures.

  • Form: Liquid
  • Format: Bottle
  • Substrate: Alkaline Phosphatase
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product LineNovaBright
Product Typeβ-Galactosidase Reporter Gene Assay System
Quantity210 mL
Shipping ConditionRoom Temperature
SubstrateAlkaline Phosphatase
Substrate TypeAP (Alkaline Phosphatase) Substrate
Unit SizeEach
Contents & Storage
Ambient for shipping. Store the product at 4°C.