Invitrogen™

Tropifluor™ PVDF Membrane, 0.45 µm pore size, 15 cm X 15 cm

Catalog number: T2234
Invitrogen™

Tropifluor™ PVDF Membrane, 0.45 µm pore size, 15 cm X 15 cm

Catalog number: T2234
Catalog Number
T2234
Unit Size
5 sheets
Price (USD)
215.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T22345 sheets
215.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
We have carefully selected and rigorously tested the membrane to provide optimized results in western blotting applications with dioxetane-based substrate solutions (CSPD® or CDP-Star® substrates).
• Our Tropifluor™ PVDF transfer membrane provides the high binding and retention capacity that characterizes PVDF (polyvinylidene fluoride) membranes.
• We have carefully selected and rigorously tested the membrane to provide optimized results in western blotting applications with dioxetane-based substrate solutions (CSPD® or CDP-Star® substrates).
• PVDF membrane provides high mechanical strength that permits convenient handling and withstands multiple re-probing.

Extend Your Detection Limits
The low background and resulting superior signal-to-noise performance obtained with this membrane enables picogram detection levels of proteins in immunoblotting applications. We have selected the Tropifluor™ membrane to yield low background levels with the use of Tropix chemiluminescent substrates and secondary antibody conjugates, enabling ultrasensitive detection. We recommend this product for all immunoblotting applications where sensitivity is important. The membrane has a pore size of 0.45 microns and a binding capacity of 125 µg of protein⁄cm2.

Products described herein are guaranteed for 12 months from the date of purchase.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
5 sheet(s)
Shipping Condition
Room Temperature
Dimensions (LxW)
15 cm x 15 cm
Format
Sheet
Membrane
PVDF Membrane
Pore Size
0.45 micron, 0.45 micron
Product Line
Tropifluor™

Contents & Storage

Membranes may be stored at room temperature.

Figures

Documents & Downloads

Certificates

Frequently asked questions (FAQs)

Citations & References