Galacton-Star® Substrate (50X Concentrate)

Catalog number: T2264

Molecular Probes™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Galacton-Star® Substrate is a concentrated chemiluminescent substrate for E.coli β-galactosidase assays.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.


Form: Liquid
Format: Bottle(s)
Substrate: Beta-Galactosidase
Product Size: 2 mL
Substrate Type: Beta-Gal Substrate
Shipping Condition: Room Temperature

Contents & storage

Store the product at 4°C.


Manuals & protocols