Invitrogen™

CDP-Star™ Substrate (12.5 mM Concentrate)

Back to top

Description

This concentrated CDP-Star® Substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CDP-Star® chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: CDP-Star®, NovaBright™
Shipping Condition: Room Temperature
Substrate: Alkaline Phosphatase
Substrate Type: AP (Alkaline Phosphatase) Substrate
Format: Bottle(s)
Quantity: 1mL

Contents & storage

Store the product at 4°C.