cAMP-Screen™ Assay/Lysis Buffer
Invitrogen™

cAMP-Screen™ Assay/Lysis Buffer

For use with the cAMP-Screen or cAmp-Screen Direct™ Immunoassay System.
Have Questions?
Catalog NumberQuantity
T232765 mL
Catalog number T2327
Price (USD)
100.00
Each
Quantity:
65 mL
Request bulk or custom format
Price (USD)
100.00
Each

This Assay/Lysis Buffer is suitable for use with the cAMP-Screen™ or cAmp-Screen Direct™ Immunoassay System.

For Research Use Only. Not for use in diagnostics procedures.

  • Quantity: 65 mL
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)cAmp-Screen Direct™ Immunoassay System., cAMP-Screen™ Immunoassay System
FormLiquid
Product LinecAMP-Screen
Shipping ConditionRoom Temperature
Product TypeELISA Assay/Lysis Buffer
Quantity65 mL
Unit SizeEach
Contents & Storage
Store the product at 4°C.