Invitrogen™

Texas Red™ C2-Dichlorotriazine

Catalog number: T30200
Invitrogen™

Texas Red™ C2-Dichlorotriazine

Catalog number: T30200
Catalog Number
T30200
also known as T-30200
Unit Size
5 mg
Price (USD)
304.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T30200
also known as T-30200
5 mg
304.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Texas Red C2-dichlorotriazine is a reactive dye with absorption/emission maxima of ∼588/601 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Alcohol, Amine
Shipping Condition
Room Temperature
Label Type
Classic Dyes
Label or Dye
Texas Red™
Product Line
Texas Red™
Reactive Moiety
Dichlorotriazine

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

Frequently asked questions (FAQs)

Citations & References