Tubulin Tracker™ Green (Oregon Green™ 488 Taxol, Bis-Acetate), for live-cell imaging
Tubulin Tracker Green (Oregon Green 488 Taxol, bis-acetate) provides green-fluorescent staining of polymerized tubulin in live cells. Tubulin Tracker GreenRead more
Have Questions?
Catalog Number
Quantity
T34075
1 set
Catalog number T34075
Price (USD)
568.10
Special offer
Online exclusive
Ends: 27-Jun-2026
598.00
Save 29.90 (5%)
1 set
Quantity:
1 set
Price (USD)
568.10
Special offer
Online exclusive
Ends: 27-Jun-2026
598.00
Save 29.90 (5%)
1 set
Tubulin Tracker Green (Oregon Green 488 Taxol, bis-acetate) provides green-fluorescent staining of polymerized tubulin in live cells. Tubulin Tracker Green is an uncharged, nonfluorescent compound that easily passes through the plasma membrane of live cells. Once inside the cell, the lipophilic blocking group is cleaved by non-specific esterases, resulting in a green-fluorescent, charged form.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeOregon Green™ 488
Product LineOregon Green
Quantity1 set
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product TypeTubulin Tracker
Sub Cellular LocalizationCytoskeleton, Tubulin
Unit Size1 set
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
What is the difference between Tubulin Tracker Green (Cat No. T34078) and Tubulin Tracker Green, for live cell imaging (Cat. No. T34075)?
Both products include the same Tubulin Tracker Green reagent. The difference is in the packaging and amount of Tubulin Tracker Green reagent contained. Tubulin Tracker Green, for live cell imaging (Cat. No. T34075) includes enough Tubulin Tracker Green reagent for 300 slides whereas Tubulin Tracker Green (Cat No. T34078) contains enough Tubulin Tracker Green reagent for 60 slides. Also, Tubulin Tracker Green, for live cell imaging (Cat. No. T34075) does not include Probenecid.
With Tubulin Tracker Deep Red and Tubulin Tracker Green, can I fix samples after labeling?
No. The labeling is not retained upon fixation. Imaging of the samples should be done only on live cells. To limit photobleaching while viewing live cells, use ProLong Live Antifade Reagent, for live cell imaging (Cat No. P36974 and P36975).
How is Docetaxel in Tubulin Tracker Deep Red different from Taxol in Tubulin Tracker Green? Are there any differences in the binding or cytotoxicity between the two reagents?
Docetaxel has a slightly higher binding affinity than Taxol and exhibits longer retention time due to better uptake and slower efflux, but for use in labeling cultured cells, these differences should be minimal. More information comparing the properties of these two reagents is provided in this publication: https://www.ncbi.nlm.nih.gov/pubmed/1671606 https://www.cell.com/fulltext/S1074-5521(05)00301-7
PKC-induced intracellular trafficking of Ca(V)2 precedes its rapid recruitment to the plasma membrane.
Authors:Zhang Y, Helm JS, Senatore A, Spafford JD, Kaczmarek LK, Jonas EA,
Journal:J Neurosci
PubMed ID:18322103
'Activation of protein kinase C (PKC) potentiates secretion in Aplysia peptidergic neurons, in part by inducing new sites for peptide release at growth cone terminals. The mechanisms by which ion channels are trafficked to such sites are, however, not well understood. We now show that PKC activation rapidly recruits new ... More
Modulation of hippocampal calcium signalling and plasticity by serine/threonine protein phosphatases.
Authors:Koss DJ, Hindley KP, Riedel G, Platt B
Journal:J Neurochem
PubMed ID:17442047
'Kinases and phosphatases act antagonistically to maintain physiological phosphorylation/dephosphorylation at numerous intracellular sites critical for neuronal signalling. In this study, it was found that inhibition of serine/threonine phosphatases by exposure of hippocampal slices to okadaic acid (OA) or cantharidin (CA; 100 nmol/L) for 2 h resulted in reduced basal synaptic ... More
Redundant mechanisms recruit actin into the contractile ring in silkworm spermatocytes.
Authors:Chen W, Foss M, Tseng KF, Zhang D,
Journal:PLoS Biol
PubMed ID:18767903
'Cytokinesis is powered by the contraction of actomyosin filaments within the newly assembled contractile ring. Microtubules are a spindle component that is essential for the induction of cytokinesis. This induction could use central spindle and/or astral microtubules to stimulate cortical contraction around the spindle equator (equatorial stimulation). Alternatively, or in ... More
2-(Naphthalene-1-yl)-6-pyrrolidinyl-4-quinazolinone inhibits skin cancer M21 cell proliferation through aberrant expression of microtubules and the cell cycle.
Authors:Wu YC, Hour MJ, Leung WC, Wu CY, Liu WZ, Chang YH, Lee HZ,
Journal:J Pharmacol Exp Ther
PubMed ID:21652781
'Microtubules are a proven target for anticancer drug development because they are critical for mitotic spindle formation and the separation of chromosomes at mitosis. 2-(Naphthalene-1-yl)-6-pyrrolidinyl-4-quinazolinone (HL66) induced cell death with the large cells and multiple micronuclei in M21 skin cancer cells. We demonstrated that HL66-induced cell death is caspase-independent and ... More
Metalloprotease meprin beta generates nontoxic N-terminal amyloid precursor protein fragments in vivo.
Authors:Jefferson T, Cauševic M, auf dem Keller U, Schilling O, Isbert S, Geyer R, Maier W, Tschickardt S, Jumpertz T, Weggen S, Bond JS, Overall CM, Pietrzik CU, Becker-Pauly C,
Journal:J Biol Chem
PubMed ID:21646356
'Identification of physiologically relevant substrates is still the most challenging part in protease research for understanding the biological activity of these enzymes. The zinc-dependent metalloprotease meprin ß is known to be expressed in many tissues with functions in health and disease. Here, we demonstrate unique interactions between meprin ß and ... More