Thermo Scientific™
Lab Vision™ Avidin biotin Blocking Solution
 Related applications:
Anatomical Pathology
Block nonspecific staining due to endogenous biotin in immunohistochemical staining with Thermo Scientific™ Lab Vision™ Avidin Biotin Blocking Solution
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Blocks nonspecific staining due to endogenous biotin during the IHC detection of antigens
Storage Conditions: Store between 4° to 6°C. Components stable for up to 18 months.
For Research Use Only
Immunohistochemical Staining Protocol:
1. Perform the A-B blocking step prior to the primary antibody. If a protein block is used before the primary, add the A-B block prior to the protein-blocking step.
2. Incubate tissue sections with Avidin Block for 10-15 minutes.
3. Wash two times with buffer.
4. Incubate tissue sections with Biotin Block for 10-15 minutes.
5. Wash two times with buffer.
6. Apply primary antibody to the tissue section.
General References:
Wood GS and Warnke R. (1981). J Histochem Cytochem. 10,1196-204.
Documents
Manuals & protocols