Thermo Scientific™
Lab Vision™ Fast-Red Substrate System
Related applications:
Clinical
Employ a widely used chromogen for immunohistochemical staining and immunoblotting Thermo Scientific™ Lab Vision™ Fast-Red Substrate System.
Back to top
In the presence of alkaline phosphatase enzyme, Fast-Red produces a bright red precipitate that is soluble in alcohol.
Includes:
TA-060-AF: 12 Fast-Red Tablets (TA-012-AFC), 60mL Naphthol Phosphate Substrate (TA-060-AFS)
TA-125-AF: 25 Fast-Red Tablets (TA-025-AFC), 125mL Naphthol Phosphate Substrate (TA-125-AFS)
Staining Protocol: 1. Add one Fast-Red tablet to 5mL of Naphthol Phosphate Substrate and shake until fully dissolved.
2. Apply solution to tissue section.
3. Incubate tissue section for 10-25 minutes.
4. Rinse tissue, counterstain, and cover slip using an aqueous mounting media.