Thermo Scientific™
Lab Vision™ Hydrogen Peroxide Block
 Related applications:
Anatomical Pathology
Quench endogenous peroxidase that can cause nonspecific background staining with the Thermo Scientific™ Lab Vision™ Hydrogen Peroxide Block.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Formulated for use with immunohistochemical procedures.
  • Peroxidase within red blood cells may be difficult to quench entirely.
For In Vitro Diagnostic use
Staining Protocol:
Hydrogen Peroxide Block may be applied after the sections have been deparaffinized and rehydrated. The application must occur prior to the primary antibody incubation. Incubate tissue section for 10-15 minutes at room temperature or 37°C. After incubation, rinse twice in buffer. For bulk staining, pour Hydrogen Peroxide Block in a covered staining tray and dip slides for 5-10 minutes. Replace with fresh Hydrogen Peroxide Block after 5-10 uses. This step can be performed at the time of deparaffinization.
For In Vitro Diagnostic use
Staining Protocol:
Hydrogen Peroxide Block may be applied after the sections have been deparaffinized and rehydrated. The application must occur prior to the primary antibody incubation. Incubate tissue section for 10-15 minutes at room temperature or 37°C. After incubation, rinse twice in buffer. For bulk staining, pour Hydrogen Peroxide Block in a covered staining tray and dip slides for 5-10 minutes. Replace with fresh Hydrogen Peroxide Block after 5-10 uses. This step can be performed at the time of deparaffinization.
Documents
Manuals & protocols