Thermo Scientific™
Lab Vision™ Trypsin (Enzymatic Pretreatment)
 Related applications:
Anatomical Pathology
Take advantage of the digestive properties of Thermo Scientific™ Lab Vision™ Trypspin (Enzymatic Pretreatment) for immunohistochemical procedures.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Supplied as a two-component system
Storage Conditions: Store at 2°-8°C. Stable for 18 months.
Includes: TA-015-TR: 15mL (Buffer) (TA-015-TRB), 5mL (Trypsin concentrate) (TA-005-TRC)
TA-125-TR: 125mL (Buffer) (TA-125-TRB), 30mL (Trypsin concentrate) (TA-030-TRC)
Procedure:
1. Combine Trypsin concentrate with buffer in 1:3 (one part to three parts) for a working solution of 0.125% Trypsin solution.
2. Incubate the Trypsin mixture on the tissue section for 10 minutes at 37°C.
3. Rinse with cool buffer solution to stop the digestive action, and proceed with remainder of immunohistochemical protocol (i.e., application of the primary antibody).
Documents
Manuals & protocols