Thermo Scientific™
Lab Vision™ UltraMount
 Related applications:
Anatomical Pathology
Cover tissue sections and cell preparations immunohistochemically stained with chromogens with Thermo Scientific™ Lab Vision™ UltraMount to produce alcohol-soluble or alcohol-insoluble end products.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Heating not required prior to use
  • Optimal for use without cover strips
  • Can be used with chromogens (Fast-Red or AEC) requiring an aqueous mounting medium
Storage Conditions: Store at room temperature. Stable until date shown on label.
Documents
Manuals & protocols