Thermo Scientific™
Lab Vision™ HIER Buffer L (100X)
 Related applications:
Anatomical Pathology
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Thermo Scientific™ Lab Vision™ HIER Buffer L (100X). Dewaxed tissue sections can be processed through the HIER buffer L, prior to manual or automated staining on a Thermo Scientific autostainer.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • For in vitro diagnostic use
  • Storage Conditions: Store at room temperature
Staining Protocol: This is a 100X stock solution and must be diluted 100-fold with deionized water before use. The final solution should be pH 6.0 ± 0.2.
1. Deparaffinize and re-hydrate sections as usual.
2. Add one bottle of HIER Buffer L in to a PT Module tank; dilute to 1.5 liters with deionized water (1:100 dilution).
3. Measure and document pH with calibrated pH meter per accreditation guidelines (optional).
4. For antigen retrieval, pre-heat the PT Module (PTM) to 85°C and immerse the slides mounted on the Autostainer rack into the PT Module tank.
5. Pre-warm the buffer to 98°C and hold for 20 min.
6. Cool slides in the PT Module to 85°C. Cool further to RT in TBST buffer for at least 5 min. Ensure slides remain moist.
7. Keep all the slides in buffer until the slides are loaded on to the Autostainer.
Documents
Manuals & protocols