Product Image
Thermo Scientific™
Thermo Scientific™ QSP™ Natural Screw Cap Micro Tubes
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection great with biologics, sample preparation, storage and transportation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClean ClaimsAutoclavableSterility
TC-520-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
511-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
512-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
512-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
514-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
515-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
515-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterile
516-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
517-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
517-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterile
519-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
520-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
521-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
522-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
522-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterile
528-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
529-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterile
520-GRDS-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterile
529-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterile
TC-512-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
TC-515-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
TC-515-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
TC-517-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
TC-517-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
TC-522-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
TC-522-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
TC-529-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
TC-529-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterile
Catalog number TC-520-GRD-Q
Price (CLP)
-
Clean Claims:
RNase/DNase, DNA, pyrogen and endotoxin free
Autoclavable:
Not Autoclavable
Sterility:
Non-sterile
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes.
Packaged in bulk bags and non-sterile clean claims are; RNase/DNase, DNA, pyrogen and endotoxin free.
  • Screw Cap micro tube volume range of 0.5, 1.5 and 2.0ml and are ideal for sample preparation, storage and transportation
  • Conical bottom or self standing offered with knurls in ultra clear natural or UV protective amber color
  • Tether caps and standard non tether caps are available in a rainbow of color options
Specifications
Clean ClaimsRNase/DNase, DNA, pyrogen and endotoxin free
AutoclavableNot Autoclavable
ColorClear
GraduatedYes
MaterialPolypropylene
SterilityNon-sterile
Unit SizeEach