2.0mL Graduated, free-standing & attached TC with O-Ring, Sterile
Available in conical and free standing designs, screw cap microcentrifuge tubes are available in volume capacities of 0.5mL, 1.5 mL and 2.0mL.
Have Questions?
Número de catálogo TC-520-GRDS-Q
Precio (USD)
-
Thermo Scientific™ QSP Screw Cap Microcentrifuge Tubes are available in conical and free standing designs. Natural and amber. These tubes are autoclavable, boil proof and freezable to -80°C. Find a complete volume range from 0.5mL through 2.0 mL.
Screw Cap Microcentrifuge Tube Design
Tubes are designed with conical bottoms allowing for more complete sample retrieval
Screw caps with O-rings guard against leakage by forming a positive seal against the rim of the tube
Tethered screw caps remain attached to the sample tube but can rotate completely for convenience
Tubes and caps are available in amber to protect your light sensitive samples
External ribs on the tubes allow for single-handed opening and closing when placed in racks with serrated holes
Screw cap microcentrifuge Tubes work with many automated systems
O-ring is manufactured from durable ethylene polypropylene dimonomere compound
Highest Quality
Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity and security during boiling, freezing and centrifugation
Nonsterile product is certified free of RNase, DNase, DNA and pyrogen
Sterile product is certified free of RNase, DNase, DNA, endotoxin, pyrogen and PCR inhibitors
Color/Packaging Options
Sterile product is packaged with the caps screwed onto the tubes
Nonsterile tubes are packaged in individually sealed bags of 500 to protect against cross contamination
Standard Screw Cap Color options are: Blue, Green, Orange, Red, Yellow and Mixed to aid with visual identification of samples
Recommended For: Daily laboratory usage including centrifugation and short term sample storage
Especificaciones
Estilo de tapónScrew cap, tethered w/o-ring
Certificaciones/conformidadFree of Rnase, Dnase, DNA, Pyrogen,PCR Inhibtor,ATP