Arktik™ Thermal Cycler, Arktik base unit without gradient - FAQs
Arktik™ Thermal Cycler, Arktik base unit without gradient - FAQs
View additional product information for Arktik™ Thermal Cycler, Arktik base unit without gradient - FAQs (TCA0002)
2 product FAQs found
Does the Arktik Thermal Cycler come with a heat block?
The Arktik thermal cycler has interchangeable blocks so the purchase of a heated block is required for operation of the Arktik thermal cycler.
Is a gradient block available with Thermo Scientific Arktik Thermal Cycler?
A 30 degrees C temperature gradient is available for the Arktik 96- and 384-well blocks*. *Not available in US, Germany or Japan