Does the Arktik Thermal Cycler come with a heat block?
The Arktik thermal cycler has interchangeable blocks so the purchase of a heated block is required for operation of the Arktik thermal cycler.
Search
View additional product information for Arktik™ Thermal Cycler, 96-well single block - FAQs (TCA0096)
2 product FAQs found