What is the temperature range of the PikoReal instrument? How accurate and uniform is the block temperature?
The PikoReal thermal cycler can operate in the temperature range of 4-99.9 degrees C. Its temperature accuracy is ±0.2 degrees C and temperature uniformity is ±0.3 degrees C at 95 degrees C, ±0.15 degrees C at 60 degrees C and ±0.2 degrees C at 72 degrees C.