TDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
TDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
View additional product information for TDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs (TDE40040LARLN, TDE50040FDRLN, TDE60040LARLN, TDE60040LDRLN, TDE40040LDRLN, TDE40040FDRLN, TDE60040FDRLN, TDE50040LARLN, TDE50040LDRLN)