TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
View additional product information for TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs (TDE40086LARLN)
2 product FAQs found
What are the electrical requirements for the TDE Series Ultra-Low Temperature Freezer (Cat. No. TDE40086LARLN)?
Here are the electrical requirements for the TDE Series Ultra-Low Temperature Freezer (Cat. No. TDE40086LARLN):
115 V, 60 Hz