TDE Series -40°C Ultra-Low Temperature Freezers - FAQs
TDE Series -40°C Ultra-Low Temperature Freezers - FAQs
View additional product information for TDE Series -40°C Ultra-Low Temperature Freezer - FAQs (TDE50040FA, TDE50040LV, TDE50040LD, TDE50040LA, TDE50040FV, TDE50040FD)