5G Terra Core Soil Sampler
Thermo Scientific™

5G Terra Core Soil Sampler

Save time with this no fuss, no mess disposable transfer tool for soil sampling.
Have Questions?
Catalog NumberProduct Type
TERRACORE5G
also known as TERRA CORE 5G
Soil Sampler
Catalog number TERRACORE5G
also known as TERRA CORE 5G
Price (MXN)
-
Product Type:
Soil Sampler
Save time with this no fuss, no mess disposable transfer tool. Thermo Scientific™ 5G Terra Core Soil Sampler is a cost effective tool for soil sampling as described in USEPA SW-846 Method 5035. The Terra Core soil sampler delivers an approximate 5g sample quickly and conveniently into a 40mL VOA vial for in-field preservation. Vial threads stay free of debris and and volatile target analytes are better maintained by minimal exposure and handling than manual spatula methods.
  • Rugged all-in-one design eliminates the need for a separate handle
  • More convenient than make-your-own cut-off syringes
  • Simple operation makes sampling easy and effective
  • Minimizes sample handling to reduce target analyte loss
Specifications
Certifications/ComplianceNone
DescriptionSoil Sampler 5g
DisposableSingle-use
Height (English)1 in.
Height (Metric)25.4 mm
Product TypeSoil Sampler
ColorNatural
MaterialPlastic
Unit SizeCase of 100