QSP Specialty Pipette Tips
Thermo Scientific™
QSP Specialty Pipette Tips
Specialty pipette tips designed for specific applications making it easier to concentrate on what is important—your research.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명부피(미터법)팁 스타일멸균제품 사이즈
TF118-200-QQSP 200 Genomic, Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
103-QQSP 10 Gel, Non-Filtered10 μLGel LoadingNon-sterileCase of 10000
103-QPK
103-Q으로도 사용됨
QSP 10 Gel, Non-Filtered10 μLGel LoadingNon-sterilePack of 1000
118-N-QQSP 200 Genomic/Wide Orifice, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 10000
118-N-QPK
118-N-Q으로도 사용됨
QSP 200 Genomic/Wide Orifice, Non-Filtered200 μLGenomic/Wide OrificeNon-sterilePack of 1000
T118R-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 4800
T118R-QPK
T118R-Q으로도 사용됨
QSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeNon-sterilePack of 960
T118RS-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
T118RS-QPK
T118RS-Q으로도 사용됨
QSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeSterilePack of 960
TF118-200-QPK
TF118-200-Q으로도 사용됨
QSP 200 Genomic, Filtered200 μLGenomic/Wide OrificeSterilePack of 960
112NXLG-QQSP 1000 Genomic Wide Orifice, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 10000
112NXLG-QPK
112NXLG-Q으로도 사용됨
QSP 1000 Genomic Wide Orifice, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterilePack of 1000
T112NXLGR-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 4800
T112NXLGR-QPK
T112NXLGR-Q으로도 사용됨
QSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterilePack of 960
T112NXLGRS-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
T112NXLGRS-QPK
T112NXLGRS-Q으로도 사용됨
QSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeSterilePack of 960
TF112G-1000-QQSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
TF112G-1000-QPK
TF112G-1000-Q으로도 사용됨
QSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterilePack of 960
카탈로그 번호 TF118-200-Q
제품 가격(KRW)
500,000
Case of 4800
설명:
QSP 200 Genomic, Filtered
부피(미터법):
200 μL
팁 스타일:
Genomic/Wide Orifice
멸균:
Sterile
Unit Size:
Case of 4800
제품 가격(KRW)
500,000
Case of 4800
Thermo Scientific™ QSP specialty pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Specialty tips are crafted for specific applications and are designed to make your work more efficient and reproducible.
Gel Loading Specialty Tips
Loading acrylamide and agarose gels with standard pipette tips is a lengthy and tedious process. Avoid damaging your gel by choosing a specialty tip that offers a longer taper with a thinner distal end that is designed for these applications.
Genomic Wide Orifice Specialty Tips:
A problem posed when working with viscous liquids like tween or thick cell lysates is pipetting reliability due to the inability to draw and dispense fluid in a uniform manner. Our large, wide orifice tips are much wider than standard tips improving capability to draw and dispense in a consistent and uniform manner.
Extended Length Specialty Tips:
Extended length tips help prevent contamination by keeping the pipette shaft from entering the sample vessel. Extended length tips can provide laboratory solutions for dispensing samples into hard to reach vessels were a standard tip just cant reach.
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
사양
클린 클레임DNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
설명QSP 200 Genomic, Filtered
길이(미터법)5cm
색상Natural
필터링됨Filtered
용도 (장비)Universal Fit
형식Hinged Rack
물질Virgin Polypropylene
제품라인QSP
멸균Sterile
팁 스타일Genomic/Wide Orifice
부피(미터법)200 μL
Unit SizeCase of 4800