Low Retention Filtered Pipette Tips
Thermo Scientific™

Low Retention Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleUnit Size
TFLR102-10-Q10 μLExtended LengthCase of 4800
TFLR102-10-QPK
also known as TFLR102-10-Q
10 μLExtended LengthPack of 960
TFLR1121000-Q1000 μLExtended LengthCase of 4800
TFLR1121000-QPK
also known as TFLR1121000-Q
1000 μLExtended LengthPack of 960
TFLR113-100-Q100 μLBeveledCase of 4800
TFLR113-100-QPK
also known as TFLR113-100-Q
100 μLBeveledPack of 960
TFLR113-20-Q20 μLBeveledCase of 4800
TFLR113-20-QPK
also known as TFLR113-20-Q
20 μLBeveledPack of 960
TFLR140-200-Q200 μLMicro PointCase of 4800
TFLR140-200-QPK
also known as TFLR140-200-Q
200 μLMicro PointPack of 960
TFLR140-300-Q300 μLMicro PointCase of 4800
TFLR140-300-QPK
also known as TFLR140-300-Q
300 μLMicro PointPack of 960
Catalog number TFLR102-10-Q
Price (KRW)
542,000
Case of 4800
Volume (Metric):
10 μL
Tip Style:
Extended Length
Unit Size:
Case of 4800
Price (KRW)
542,000
Case of 4800
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.

Featuring:

  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
GraduatedYes
Graduations2, 5, 10, 20 μL
Length (Metric)4.6 cm
ModelQSP 10XL
ColorNatural
FilteredFiltered
For Use With (Equipment)Universal Fit
FormatHinged Rack
Product LineQSP
SterilitySterile
Tip StyleExtended Length
Volume (Metric)10 μL
Unit SizeCase of 4800