Low Retention Filtered Pipette Tips
Thermo Scientific™

Low Retention Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
TFLR140-200-Q200 μLMicro Point
TFLR102-10-Q10 μLExtended Length
TFLR1121000-Q1000 μLExtended Length
TFLR113-100-Q100 μLBeveled
TFLR113-20-Q20 μLBeveled
TFLR140-300-Q300 μLMicro Point
Catalog number TFLR140-200-Q
Price (BRL)
-
Volume (Metric):
200 μL
Tip Style:
Micro Point
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.

Featuring:

  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
GraduatedYes
Green FeaturesACT Ecolabel (Silver)
Length (Metric)5.8 cm
ModelQSP 200
ColorNatural
FilteredFiltered
For Use With (Equipment)Universal Fit
FormatHinged Rack
Product LineQSP
SterilitySterile
Tip StyleMicro Point
Volume (Metric)200 μL
Unit SizeCase of 4800