iCAP™ 7000 Series ICP-OES eLearning Courses
Thermo Scientific™
iCAP™ 7000 Series ICP-OES eLearning Courses
The iCAP™ 7000 Series ICP-OES eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCourse Topic
TICAP7MTSMantenimiento por parte del usuario y solución de problemas
TICAP7FTHTeoría fundacional
TICAP7BOPOperaciones básicas
Número de catálogo TICAP7MTS
Precio (EUR)
-
Course Topic:
Mantenimiento por parte del usuario y solución de problemas
These eLearning courses are an introduction to the basic principles of ICP-OES and how the iCAP 7000 Series instruments use this technology.
The iCAP™ 7000 Series Basic Operations eLearning course
will provide you with the basic training on the theory, operation, and general maintenance of your instrument. Included in the e-Learning course are the following e-learning modules:
iCAP™ 7000+ ICP-OES:
  • Peristaltic Pump and Pump Tubing Maintenance
  • Concentric Glass Nebulizer
  • Glass Cyclonic Spray Chamber
  • Plasma Torch Assembly
  • Purged Optical Path (POP) Window Maintenance
  • ThermoFlex 900 Recirculating Chiller
  • Thermo Scientific™ Qtegra™ ISDS Software: Introduction to the Qtegra Software
  • Thermo Scientific™ Qtegra™ ISDS Software: Creating a LabBook
  • Thermo Scientific™ Qtegra™ ISDS Software: Reviewing and Exporting Data and Reports
  • iCAP 7000+ ICP-OES: Creating Calibration Standards Volumetrically
  • iCAP 7000+ ICP-OES: Startup and Shutdown Procedures
Especificaciones
Access Availablity30 days upon end user first access to the course
Course TopicMantenimiento por parte del usuario y solución de problemas
FormatoSelf-paced eLearning
IdiomasEnglish
Course LevelRoutine
Duración60 to 120 min.
Para utilizar con (equipo)iCAP 7000 Series ICP-OES
Línea de productosThermo Scientific
Tipo de productoAtomic Spectroscopy
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach