Dionex™ Inuvion™ IC System eLearning Courses
Thermo Scientific™

Dionex™ Inuvion™ IC System eLearning Courses

The eLearning courses for the Dionex™ Inuvion™ IC System provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse LevelDuration
TINUMTSUser Maintenance and TroubleshootingRoutine60 to 120 min.
TINUFTHFoundational TheoryFoundation30 to 60 min.
TINUBOPBasic OperationsRoutine60 to 120 min.
Catalog number TINUMTS
Price (MXN)
-
Course Topic:
User Maintenance and Troubleshooting
Course Level:
Routine
Duration:
60 to 120 min.

The eLearning courses for the Dionex™ Inuvion™ IC System provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses that can cover different topics such as foundational theory, basic operations principles, and/or user maintenance and troubleshooting.

The Dionex™ Inuvion™ IC System Foundational Theory eLearning course
Is an introduction to the general principles of ion chromatography.This course provides a review of the basic principles of chromatography and the history of the development of ion chromatography (IC) technology. The standard components of the IC system and how these facilitate the successful separation of analytes will be covered. Additional concepts discussed include:

  • What is ion chromatography?
  • A detailed description of the ion exchange process
  • Effects of mobile and stationary phases
  • Basic overview of Reagent-Free™ Ion Chromatography (RFIC™) technology
  • IC columns and their factors affecting peak resolution
  • Principles of suppressed conductivity detection

The Dionex™ Inuvion™ IC System Basic Operations with Chromeleon™ eLearning course
Provides basic information regarding the routine preparation steps required before running samples using a Thermo Scientific™ Dionex™ Inuvion™ IC System with conductivity detection. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. This course will also provide a detailed step-by-step instruction checklist to install a new DRS 600 suppressor on your ion chromatography system.

Some of the topics covered include:

  • Filling the eluent reservoirs
  • Priming the ion chromatograph fluidics
  • Overview, installation, operation, and troubleshooting guidelines for Reagent Free Ion Chromatography (RFIC) consumables often used in standard IC systems
    • EGC: Eluent Generator Cartridge
    • Electrolytic Suppressor
  • Manual instrument startup
  • Operating status verification
  • Loading samples in the sampler
  • Creating sequences and methods
  • Processing samples

The Dionex™ Inuvion™ IC System Basic Operations with Chromeleon™ eLearning course
Provides basic information regarding the routine preparation steps required before running samples using a Thermo Scientific™ Dionex™ Inuvion™ IC System with conductivity detection. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. This course will also provide a detailed step-by-step instruction checklist to install a new DRS 600 suppressor on your ion chromatography system.

Some of the topics covered include:

  • Filling the eluent reservoirs
  • Priming the ion chromatograph fluidics
  • Overview, installation, operation, and troubleshooting guidelines for Reagent Free Ion Chromatography (RFIC) consumables often used in standard IC systems
    • EGC: Eluent Generator Cartridge
    • Electrolytic Suppressor
  • Manual instrument startup
  • Operating status verification
  • Loading samples in the sampler
  • Creating sequences & methods
  • Processing samples

The Dionex™ Inuvion™ IC System User Maintenance and Troubleshooting eLearning course
Describes routine user maintenance procedures for high-pressure valves and pumps installed in Thermo Scientific™ Dionex™ Inuvion™ IC Systems. It provides a review of an overall description of an Ion Chromatography System setup with conductivity detection and discusses its vital signs. Some of the topics covered include:

  • Detailed instructions on the routine maintenance procedures for high-pressure valves:
    • Rotor and stator face seal replacement
    • Stator care tips
  • Detailed instructions to perform the following standard-flow pump maintenance procedures:
    • Removing and remounting pump heads
    • Cleaning and inspecting pistons
    • Replacing check valves, main piston seals, and rear seal wash seals
  • Troubleshooting tips on the most probable symptoms/problems that will help you quickly resume operation to generate accurate, reliable data.
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicUser Maintenance and Troubleshooting
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration60 to 120 min.
For Use With (Equipment)Dionex Inuvion
Product LineThermo Scientific
Product TypeChromatography
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach