Thermo Scientific™
UltraVision™ Quanto Mouse on Mouse
 Related applications:
Anatomical Pathology
Provide strong staining while reducing mouse on mouse background all with a simple routine procedure using the Thermo Scientific™ UltraVision™ Quanto Mouse on Mouse HRP IHC detection system. Makes optimal results routine with mouse antibodies on mouse tissue.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Thermo Scientific UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely. UltraVision Quanto Makes Optimal Results Routine by providing:
  • Strong Staining
  • Reduced Mouse on Mouse Background
  • Simple Biotin Free Polymer Procedure
  • For use with a DAB or other HRP Chromogen; For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL)
Documents
Manuals & protocols