QSP Low Retention Non-Filtered Pipette Tips
Thermo Scientific™

QSP Low Retention Non-Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFormatSterility
TLR112NXL-Q1000 μLExtended LengthBulkNon-sterile
TLR102-Q10 μLExtended LengthBulkNon-sterile
TLR102RS-Q10 μLExtended LengthHinged RackSterile
TLR102RL-Q10 μLExtended LengthRefillNon-sterile
TLR102RLS-Q10 μLExtended LengthRefillSterile
TLR070-Q200 μLBeveledBulkNon-sterile
TLR070R-Q200 μLBeveledHinged RackNon-sterile
TLR070RS-Q200 μLBeveledHinged RackSterile
TLR070RL-Q200 μLBeveledRefillNon-sterile
TLR070RLS-Q200 μLBeveledRefillSterile
TLR106-Q300 μLMicro PointBulkNon-sterile
TLR106R-Q300 μLMicro PointHinged RackNon-sterile
TLR106RS-Q300 μLMicro PointHinged RackSterile
TLR106RL-Q300 μLMicro PointRefillNon-sterile
TLR106RLS-Q300 μLMicro PointRefillSterile
TLR112NXLR-Q1000 μLExtended LengthHinged RackNon-sterile
TLR112NXLRS-Q1000 μLExtended LengthHinged RackSterile
TLR112NXLRL-Q1000 μLExtended LengthRefillNon-sterile
TLR112NXRLS-Q1000 μLExtended LengthRefillSterile
Catalog number TLR112NXL-Q
Price (CLP)
-
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Format:
Bulk
Sterility:
Non-sterile
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.

Featuring:

  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 1000, Non-Filtered
Length (Metric)8.9 cm
ModelQSP 1000
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatBulk
Product LineQSP
SterilityNon-sterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizeCase of 10000