QSP Low Retention Filtered And Non-Filtered Pipette Tips
Thermo Scientific™

QSP Low Retention Filtered And Non-Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleFiltered
TLR112NXLRL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR102-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102RS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102RL-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102RLS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TFLR113-20-Q20 μLBeveledFiltered
TFLR113-100-Q100 μLBeveledFiltered
TLR070-QQSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070R-QQSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RS-QQSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RL-QQSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RLS-QQSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR106-QQSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106R-QQSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RS-QQSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RL-QQSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RLS-QQSP 300, Non-Filtered300 μLMicroPointNon-filtered
TFLR140-200-Q200 μLMicroPointFiltered
TFLR140-300-Q300 μLMicroPointFiltered
TLR112NXL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR112NXLR-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR112NXLRS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR112NXRLS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TFLR1121000-Q1000 μLExtended LengthFiltered
TFLR102-10-Q10 μLExtended LengthFiltered
Catalog number TLR112NXLRL-Q
Price (TWD)
-
Description:
QSP 1000, Non-Filtered
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Filtered:
Non-filtered
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.


Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 1000, Non-Filtered
Length (Metric)8.9cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatRefill
Product LineQSP
SterilityNon-sterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizeCase of 4800