QSP Low Retention Filtered And Non-Filtered Pipette Tips
Thermo Scientific™

QSP Low Retention Filtered And Non-Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleFiltered
TLR112NXLRS-QPK
also known as TLR112NXLRS-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR102-QPK
also known as TLR102-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102R-QPK
also known as TLR102R-Q
QSP 10XL, Non-Filtered10 μLExtended Length, ConicalNon-filtered
TLR102RS-QPK
also known as TLR102RS-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102RL-QPK
also known as TLR102RL-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TLR102RLS-QPK
also known as TLR102RLS-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filtered
TFLR113-20-QPK
also known as TFLR113-20-Q
20 μLBeveledFiltered
TFLR113-100-QPK
also known as TFLR113-100-Q
100 μLBeveledFiltered
TLR070-QPK
also known as TLR070-Q
QSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070R-QPK
also known as TLR070R-Q
QSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RS-QPK
also known as TLR070RS-Q
QSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RL-QPK
also known as TLR070RL-Q
QSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR070RLS-QPK
also known as TLR070RLS-Q
QSP 200, Non-Filtered200 μLBeveledNon-filtered
TLR106-QPK
also known as TLR106-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106R-QPK
also known as TLR106R-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RS-QPK
also known as TLR106RS-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RL-QPK
also known as TLR106RL-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filtered
TLR106RLS-QPK
also known as TLR106RLS-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filtered
TFLR140-200-QPK
also known as TFLR140-200-Q
200 μLMicroPointFiltered
TFLR140-300-QPK
also known as TFLR140-300-Q
300 μLMicroPointFiltered
TLR112NXL-QPK
also known as TLR112NXL-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR112NXLR-QPK
also known as TLR112NXLR-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TLR112NXLRL-QPK
also known as TLR112NXLRL-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filtered
TFLR1121000-QPK
also known as TFLR1121000-Q
1000 μLExtended LengthFiltered
TFLR102-10-QPK
also known as TFLR102-10-Q
10 μLExtended LengthFiltered
Catalog number TLR112NXLRS-QPK
also known as TLR112NXLRS-Q
Price (INR)
3,504.00
Pack of 960
Description:
QSP 1000, Non-Filtered
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Filtered:
Non-filtered
Price (INR)
3,504.00
Pack of 960
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.


Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
DescriptionQSP 1000, Non-Filtered
Length (Metric)8.9cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatHinged Rack
Product LineQSP
SterilitySterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizePack of 960