VetMAX™ MAP IS900-F57 Kit
Applied Biosystems™

VetMAX™ MAP IS900-F57 Kit

Have Questions?
Catalog NumberQuantity
TMPT100 tests
Catalog number TMPT
Price (HKD)
-
Quantity:
100 tests
Note: The name of this product has changed. It was previously called LSI VetMAX Triplex M. Paratuberculosis.

The LSI VetMAX™ Triplex M. paratuberculosis Advanced Kit is a single-well triplex real-time PCR kit based on IS900 and F57 target genes, for the screening and confirmation of the presence of MAP DNA in one test run. The kit includes internal controls that help prevent false-negative results by verifying correct extraction and amplification. This kit is intended for use with cattle, sheep, and goat, and is validated for multiple sample types including feces, organs, bacterial colonies, and broth culture.
Specifications
DiseaseParatuberculosis
FormatTube
Product LineLSI VetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 tests
SpeciesCattle/Bovine
Target Organism ClassMycobacterium avium spp paratuberculosis (MAP)
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR)
Unit Size100 tests
Contents & Storage
Store in freezer (-5° to -30°C).