Thermo Scientific™

GGT Reagents

Catalog number: TR19110
Thermo Scientific™

GGT Reagents

Catalog number: TR19110
Determine quantity of gamma-glutamyl transferase (GGT) in human serum or plasma in vitro with Thermo Scientific™ GGT Reagents.
 
Catalog Number
TR19110
Unit Size
Each
Quantity
20 x 10 mL
Price (USD)
Full specifications
Certifications/ComplianceCE Marked
Quantity20 x 10 mL
CE MarkerYes
DescriptionGGT
Detectable AnalytesGGT
Linearity0 to 600U/L
Methods CompatibilityKinetic
Stability21 days (Reconstituted)
FormatLiquid
Sample TypePlasma
TypeReagent
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
TR19110Full specifications
Each20 x 10 mLRequest A Quote
Certifications/ComplianceCE Marked
Quantity20 x 10 mL
CE MarkerYes
DescriptionGGT
Detectable AnalytesGGT
Linearity0 to 600U/L
Methods CompatibilityKinetic
Stability21 days (Reconstituted)
FormatLiquid
Sample TypePlasma
TypeReagent
Unit SizeEach
Showing 1 of 1

GGT present in the serum originates primarily from the hepatobiliary system, and its activity is elevated in all forms of liver disease. Increased concentrations of the enzyme are also found in chronic alcoholism and with anticonvulsant therapy.

  • Reagents offer the laboratory 21 days reconstituted stability
For In Vitro Diagnostic Use.  

Contents & Storage

2°C to 8°C

Figures

Frequently asked questions (FAQs)

Citations & References