Product Image
Thermo Scientific™

LDH Reagents

Determine in vitro quantity of LDH in human serum in vitro with Thermo Scientific™ LDH Reagents.
Have Questions?
Catalog NumberQuantity
TR2001520 x 20 mL
Catalog number TR20015
Price (CLP)
-
Quantity:
20 x 20 mL
Determine in vitro quantity of LDH in human serum in vitro with Thermo Scientific™ LDH Reagents.

Lactate dehydrogenase (LDH) is concentrated in the liver, heart, kidney, skeletal muscle and erythrocytes. Increases are observed after myocardial infarction and in liver disease, untreated pernicious anemia, renal disease, malignancy and progressive muscular dystrophy.

  • At least five days reconstituted stability
  • For use in manual or automated systems
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionInfinity Triglyceride 2 x 125mL
LinearityUp to 10mmol/L (885mg/dL)
Methods CompatibilityEndpoint
Quantity20 x 20 mL
Sample TypePlasma, Serum
Stability18 months at 2-8°C
TypeReagent
CE MarkerYes
Detectable AnalytesLDH
Unit SizeEach